Learn More
Microscope World SCHOTT KL 2500 LED Dual Pipe Light Illuminator
SDP
Catalog No. NC2965464
Encompass_Preferred
This SCHOTT cold light source pipe light system includes a dual gooseneck light guide, 2 focusing light guide attachments, and a KL 2500 LED cold light source.Gooseneck- Dual Flexible and Obedient 600mm (23.62inch) long, tip is 6mm (0.24inch) diameter.Focusing- Focusing attachments screw onto each dual light guide.Color Temperature- 5600KLuminous Flux- Up to11002 lm light flux.Voltage- 100-240V, 50-60Hz.Light Box- Intensity control 0-100%, and filter slot with 2-position filter slider. LCD display indicates brightness / operation mode. USB connectivity for PC control.Light Box Dimensions / Weight- 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 3.6kg (7.9 lb)Certifications- CEWarranty- 2 years
Catalog No. NC2965464 Supplier Microscope World Supplier No. KL2500DUAL
This SCHOTT cold light source pipe light system includes a dual gooseneck light guide, 2 focusing light guide attachments, and a KL 2500 LED cold light source.Gooseneck- Dual Flexible and Obedient 600mm (23.62inch) long, tip is 6mm (0.24inch) diameter.Focusing- Focusing attachments screw onto each dual light guide.Color Temperature- 5600KLuminous Flux- Up to11002 lm light flux.Voltage- 100-240V, 50-60Hz.Light Box- Intensity control 0-100%, and filter slot with 2-position filter slider. LCD display indicates brightness / operation mode. USB connectivity for PC control.Light Box Dimensions / Weight- 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 3.6kg (7.9 lb)Certifications- CEWarranty- 2 years