Learn More
Microscope World SCHOTT ColdVision MC-LS Dual Pipe Illuminator
SDP
Catalog No. NC2965654
Encompass_Preferred
This SCHOTT ColdVision MC-LS LED Light Source with dual pipe lights is designed for use in stereo microscopy. The system includes the MC-LS LED light source and a dual gooseneck light guide with focusing lenses.Gooseneck- The 2-branch goosenecks measure 584mm (23inch). The tubing measures 12mm (0.47inch) in diameter, and the gooseneck tips are 16mm (0.63inch).Color Temperature- 5400K cool white.Luminous Flux- Up to 8502 lm light flux.Max Active Light Guide Ø- 13mmVoltage- 100-240V, 50/60HzLight Box- Continuous dimming, fan cooling, temperature monitoring for protection of the LED light engine, high resistance to vibration and shock.Analog and Digital Remote Control- USB, RS-232, 9-pin connector.Light Box Dimensions / Weight- 158mm x 145mm x 98mm (6.2inch x 5.7inch x 3.9inch), 2kg (4.62 lb)Certifications- ETL Approved, RoHS compliant.Warranty- 2 years
Catalog No. NC2965654 Supplier Microscope World Supplier No. MCLSDUAL
This SCHOTT ColdVision MC-LS LED Light Source with dual pipe lights is designed for use in stereo microscopy. The system includes the MC-LS LED light source and a dual gooseneck light guide with focusing lenses.Gooseneck- The 2-branch goosenecks measure 584mm (23inch). The tubing measures 12mm (0.47inch) in diameter, and the gooseneck tips are 16mm (0.63inch).Color Temperature- 5400K cool white.Luminous Flux- Up to 8502 lm light flux.Max Active Light Guide Ø- 13mmVoltage- 100-240V, 50/60HzLight Box- Continuous dimming, fan cooling, temperature monitoring for protection of the LED light engine, high resistance to vibration and shock.Analog and Digital Remote Control- USB, RS-232, 9-pin connector.Light Box Dimensions / Weight- 158mm x 145mm x 98mm (6.2inch x 5.7inch x 3.9inch), 2kg (4.62 lb)Certifications- ETL Approved, RoHS compliant.Warranty- 2 years