Learn More
Microscope World SCHOTT EasyLED Dual Spot Light Illuminator
SDP
Catalog No. NC2965629
Encompass_Preferred
The SCHOTT EasyLED spot light system has dual spot lights with an integrated controller. The light is cooled by convection and the spot lights can be switched on independently.Control Unit Dimensions- 61mm x 114mm x 107mm (2.40inch x 4.49inch x 4.21inch)Spot Light Length- 500mm (19.69inch)Color Temperature- 5600KLuminous Flux- 130 lmVoltage- 100~240V, 50-60HzWeight- 0.7kg (1.5lb)Certifications- CE, UL, PSE, UKCAWarranty- 2 years
Catalog No. NC2965629 Supplier Microscope World Supplier No. EASYLED2PIPE
The SCHOTT EasyLED spot light system has dual spot lights with an integrated controller. The light is cooled by convection and the spot lights can be switched on independently.Control Unit Dimensions- 61mm x 114mm x 107mm (2.40inch x 4.49inch x 4.21inch)Spot Light Length- 500mm (19.69inch)Color Temperature- 5600KLuminous Flux- 130 lmVoltage- 100~240V, 50-60HzWeight- 0.7kg (1.5lb)Certifications- CE, UL, PSE, UKCAWarranty- 2 years