Learn More
BINDER™ LED Light Bars
SDP
Catalog No. 08111253 Shop All BINDER Inc. Products
Change view
Click to view available options
Length (Metric):
30 cm
50 cm
90 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
08-111-253 90 cm
08-111-249 30 cm
08-111-251 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08-111-253 Supplier BINDER™ Supplier No. 80121109
Add to Cart
Add to Cart
LED light bars - Basic set consisting of 2 pieces, attachment material, control unit for max. 4 light strips, 100-240 V, 50/60 Hz.
  • 2 pieces
  • Includes attachment material
  • Control unit for max. 4 light strips
  • 100-240 V, 50/60 Hz
Specifications
Description LED light bars - Basic set 900, length 90 cm, Basic set consisting of 2 Pc., attachment material, control unit for max. 4 light strips, 100-240 V, 50/60 Hz
For Use With (Equipment) BINDER Chambers KB 400, KB 720, KB ECO720, KB ECO 1020, KBF-S 720, KBF-S 1020, KBF-S ECO 720, KBF-S ECO 1020, KBF 720, KBF 1020, , KMF 720
Lighting LED
Material Plastic
Type Lighting
Includes 4 light strips with attachment material
Length (English) 35.4 in.
Length (Metric) 90 cm
Voltage 100/240 V