Learn More
PCE AMERICAS INC Lux Meter measures up to 400000 lx.
SDP
Catalog No. NC3654892
Encompass_Preferred
"The illuminometer is used wherever a light check is necessary or recommended. The external light sensor of the illuminometer can be positioned according to requirements. The sensor cable has an extended length of approx. 1.5 metres. Different work or production projects require different lighting conditions.Measuring ranges:40 / 400 / 4000 / 40000 / 400000 luxMeasuring units: Lux or footcandlesMeasuring accuracy: ±3 %Accuracy geared towards LED white lightespecially for LEDsMIN- / MAX- / AVG-HOLD2.5 measurements per secondInternal memory: 99 measured values"
Catalog No. NC3654892 Supplier PCE AMERICAS INC Supplier No. PCELED 30
"The illuminometer is used wherever a light check is necessary or recommended. The external light sensor of the illuminometer can be positioned according to requirements. The sensor cable has an extended length of approx. 1.5 metres. Different work or production projects require different lighting conditions.Measuring ranges:40 / 400 / 4000 / 40000 / 400000 luxMeasuring units: Lux or footcandlesMeasuring accuracy: ±3 %Accuracy geared towards LED white lightespecially for LEDsMIN- / MAX- / AVG-HOLD2.5 measurements per secondInternal memory: 99 measured values"