Learn More
Caron Products And Services LED Plant Growth, 1-Door, 22 cu. ft., 600 mole, 3 Tier
Catalog No. NC2292396
Encompass_Preferred
LED Plant Growth Chamber. 22 ft3 with 600 mole/m2/s of LED light, 3 tiers, temperature control, touchscreen. 10C to 45C, 115V, 60 Hz.
Catalog No. NC2292396 Supplier Caron Products And Services Supplier No. 7312221
Add to Cart
Add to Cart
Request a Quote
LED Plant Growth Chamber. 22 ft3 with 600 mole/m2/s of LED light, 3 tiers, temperature control, touchscreen. 10C to 45C, 115V, 60 Hz.