Learn More
Caron Products And Services LED Plant Growth, 1-Door, 22 cu. ft., 150 mole, 6 Tier
Catalog No. NC2292401
Encompass_Preferred
LED Plant Growth Chamber optimized for tissue culture applications. 22 ft3 with 150 mole/m2/s light, 6 tiers, temperature control, touchscreen. 10C to 45C, 115V, 60 Hz.
This item is not returnable. View return policy
Catalog No. NC2292401 Supplier Caron Products And Services Supplier No. 7315221
Add to Cart
Add to Cart
Request a Quote
LED Plant Growth Chamber optimized for tissue culture applications. 22 ft3 with 150 mole/m2/s light, 6 tiers, temperature control, touchscreen. 10C to 45C, 115V, 60 Hz.