Learn More
World Precision Instrument LED Ring Light
SDP
Catalog No. 502539400
Encompass
  • “White” light illumination — 72 LED bulbs
  • Maximum opening 61 mm
  • Ring light is divided into four areas and each area is turned on and off separately
  • Brightness adjustable
  • ESD safe
  • Power supply AC 90-264V, 50/60 Hz, US plug only
Catalog No. 50-253-9400 Supplier World Precision Instrument Supplier No. 504134
  • “White” light illumination — 72 LED bulbs
  • Maximum opening 61 mm
  • Ring light is divided into four areas and each area is turned on and off separately
  • Brightness adjustable
  • ESD safe
  • Power supply AC 90-264V, 50/60 Hz, US plug only