Learn More
Unitron Ltd LED RING LIGHT VAR 60.5MM ID
SDP
Catalog No. NC0379705
Encompass_Preferred
Designed for the cost-conscious consumer, these light sources offer good value as users transition away from older fluorescent and halogen light sources.Features include 72 LED bulbs with 10-step variable-intensity light control, sectional lighting control that allows users to direct the light from four separate quadrants, a 60.5-mm inner diameter and color temperature of 6400 K. Lamp life is 10,000 h, and the power supply is AC 90/265 V at 50/60 Hz. Output is 12 V at 400 mA.
Catalog No. NC0379705 Supplier Unitron Ltd Supplier No. 15854
Designed for the cost-conscious consumer, these light sources offer good value as users transition away from older fluorescent and halogen light sources.Features include 72 LED bulbs with 10-step variable-intensity light control, sectional lighting control that allows users to direct the light from four separate quadrants, a 60.5-mm inner diameter and color temperature of 6400 K. Lamp life is 10,000 h, and the power supply is AC 90/265 V at 50/60 Hz. Output is 12 V at 400 mA.