Learn More
Caron Products And Services LED Tissue/Seedling, 1-Door, 22 cu. ft., 300 mole, 4 Tier
Catalog No. NC2292400
Encompass_Preferred
LED Plant Growth Chamber. 22 ft3 with 300 mole/m2/s of gBrite LED light, 4 tier, temperature control, touchscreen. 10C to 45C, 115V, 60 Hz.
This item is not returnable. View return policy
Catalog No. NC2292400 Supplier Caron Products And Services Supplier No. 7314221
Add to Cart
Add to Cart
Request a Quote
LED Plant Growth Chamber. 22 ft3 with 300 mole/m2/s of gBrite LED light, 4 tier, temperature control, touchscreen. 10C to 45C, 115V, 60 Hz.