Learn More
Medchemexpress LLC LED209 | 245342-14-7 | 98.4% | 383.49 | 200 MG
SDP
Catalog No. 5000400945
Encompass_Preferred
LED209 is a potent and orally active small molecule inhibitor of the bacterial receptor QseC and a potent prodrug with high selectivity for QseC. It inhibits the binding of signaling molecules to QseC and exhibits antibacterial activity.
  • Potent and orally active small molecule inhibitor of bacterial receptor QseC.
  • Potent prodrug with high selectivity for QseC.
  • Inhibits the binding of signaling molecules to QseC.
  • Has antibacterial activity.
  • Decreases biofilm formation in EAEC O104:H4 and multidrug-resistant clinical isolates of rUTIs.
  • Reduces L. pneumophila replication.
  • Protects mice against S. Typhimurium and F. tularensis murine infections.
Catalog No. 50-004-00945 Supplier Medchemexpress LLC Supplier No. HY19748200MG
LED209 is a potent and orally active small molecule inhibitor of the bacterial receptor QseC and a potent prodrug with high selectivity for QseC. It inhibits the binding of signaling molecules to QseC and exhibits antibacterial activity.
  • Potent and orally active small molecule inhibitor of bacterial receptor QseC.
  • Potent prodrug with high selectivity for QseC.
  • Inhibits the binding of signaling molecules to QseC.
  • Has antibacterial activity.
  • Decreases biofilm formation in EAEC O104:H4 and multidrug-resistant clinical isolates of rUTIs.
  • Reduces L. pneumophila replication.
  • Protects mice against S. Typhimurium and F. tularensis murine infections.