Learn More
Abcam Ledipasvir, 5A of hepatitis C virus inhibitor, 5MG
Catalog No. 502672635
Encompass_Preferred
MW 889 Da. Ledipasvir is an inhibitor of non-structural protein 5A of hepatitis C virus. It has potent effects against genotypes 1a, 1b, 4a, and 5a in vitro, but has lower activity against genotypes 2a and 3a. Ledipasvir can have additive to synergistic in vitro antiviral activity when combined with other agents.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2635 Supplier Abcam Supplier No. AB2315085MG
MW 889 Da. Ledipasvir is an inhibitor of non-structural protein 5A of hepatitis C virus. It has potent effects against genotypes 1a, 1b, 4a, and 5a in vitro, but has lower activity against genotypes 2a and 3a. Ledipasvir can have additive to synergistic in vitro antiviral activity when combined with other agents.
The product is subject to the following: Abcam Restricted Use Statement