Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ledipasvir (acetone) | 1441674-54-9 | MFCD28167732 | 99.9% | 947.08 g/mol | C52H60F2N8O7 | 10 MG
SDP

Catalog No. 5000288130
Encompass_Preferred

Ledipasvir (acetone) is the active form of the antiviral compound ledipasvir and a potent hepatitis C virus (HCV) NS5A inhibitor. It is supplied as a 10 mg research-scale sample with recommended storage and solubility conditions for laboratory use.

  • Shows potent NS5A inhibition with EC50 values of 34 pM (GT1a) and 4 pM (GT1b).
  • Molecular formula: C52H60F2N8O7.
  • Molecular weight: 947.08 g/mol.
  • CAS number: 1441674-54-9.
  • Purity: 99.9%.
  • Package size: 10 mg.
  • In vivo solubility ≥ 2.5 mg/mL (2.64 mM); clear solution.
  • Storage: 4°C protected from light; in solvent, -80°C (6 months) or -20°C (1 month), protect from light.

Catalog No. 50-002-88130 Supplier Medchemexpress LLC Supplier No. HY15602A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ledipasvir (acetone) is the active form of the antiviral compound ledipasvir and a potent hepatitis C virus (HCV) NS5A inhibitor. It is supplied as a 10 mg research-scale sample with recommended storage and solubility conditions for laboratory use.

  • Shows potent NS5A inhibition with EC50 values of 34 pM (GT1a) and 4 pM (GT1b).
  • Molecular formula: C52H60F2N8O7.
  • Molecular weight: 947.08 g/mol.
  • CAS number: 1441674-54-9.
  • Purity: 99.9%.
  • Package size: 10 mg.
  • In vivo solubility ≥ 2.5 mg/mL (2.64 mM); clear solution.
  • Storage: 4°C protected from light; in solvent, -80°C (6 months) or -20°C (1 month), protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.