Learn More
TARGETMOL CHEMICALS INC LEDIPASVIR ACETONE 25MG
SDP
Catalog No. 502579931
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ledipasvir (acetone) (GS-5885 acetone) is an HCV NS5A polymerase inhibitor used for the treatment of hepatitis C virus infection. purity: 100%
Catalog No. 50-257-9931 Supplier TARGETMOL CHEMICALS INC Supplier No. T420325MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ledipasvir (acetone) (GS-5885 acetone) is an HCV NS5A polymerase inhibitor used for the treatment of hepatitis C virus infection. purity: 100%