Learn More
Medchemexpress LLC HY-15602A 5mg Medchemexpress, Ledipasvir (acetone) CAS:1441674-54-9 Purity:>98%
SDP
Catalog No. 502081747
Encompass_Preferred
Medchemexpress, HY-15602A 5mg Ledipasvir (acetone) CAS:1441674-54-9 Ledipasvir acetone (GS-5885 acetone) is the active ingredient of Ledipasvir. Ledipasvir is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1747 Supplier Medchemexpress LLC Supplier No. HY15602A5MG
Medchemexpress, HY-15602A 5mg Ledipasvir (acetone) CAS:1441674-54-9 Ledipasvir acetone (GS-5885 acetone) is the active ingredient of Ledipasvir. Ledipasvir is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.