Learn More
Selleck Chemical LLC Ledipasvir acetone S4434-5mg
SDP
Catalog No. 503109331
Encompass_Preferred
Ledipasvir (GS-5885) acetone is the active ingredient of Ledipasvir Ledipasvir is a potent oral active inhibitor of the hepatitis C virus (HCV) NS5A with EC50 of 31 pM and 4 pM against GT1a and GT1b replicon respectively
Catalog No. 50-310-9331 Supplier Selleck Chemical LLC Supplier No. S44345MG
Ledipasvir (GS-5885) acetone is the active ingredient of Ledipasvir Ledipasvir is a potent oral active inhibitor of the hepatitis C virus (HCV) NS5A with EC50 of 31 pM and 4 pM against GT1a and GT1b replicon respectively