Learn More
Medchemexpress LLC HY-15602B 5mg Medchemexpress, Ledipasvir (D-tartrate) CAS:1502654-87-6 Purity:>98%
SDP
Catalog No. 502081748
Encompass_Preferred
Medchemexpress, HY-15602B 5mg Ledipasvir (D-tartrate) CAS:1502654-87-6 Ledipasvir D-tartrate is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1748 Supplier Medchemexpress LLC Supplier No. HY15602B5MG
Medchemexpress, HY-15602B 5mg Ledipasvir (D-tartrate) CAS:1502654-87-6 Ledipasvir D-tartrate is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.