Learn More
AdipoGen Ledol (1 g)
SDP
Supplier:  AdipoGen AGCN20406G001
Encompass_Preferred
Chemical. CAS: 577-27-5. Formula: C15H26O. MW: 222.4. Isolated from Ledum palustre. Toxic sesquiterpenoid. Exhibits expectorant and antitussive effect. Reported to increase blood pressure. Antifungal compound.
Catalog No. 50-205-3574