Learn More
Med Vet International LEDPortable Light Source w/ 1700mAh Charger, J1637, Each
Catalog No. 5000585701
Encompass_Preferred
LED Portable Lightsource for all Scopes • Visualization with minimal equipment • Easy illumination
Catalog No. 50-005-85701 Supplier Med Vet International Supplier No. JOR1637
LED Portable Lightsource for all Scopes • Visualization with minimal equipment • Easy illumination