Learn More
NEOGEN LEE Broth, 500 g
SDP
Catalog No. 502015720
Encompass_Preferred
Listeria Express Enrichment Broth (LEE Broth) is a selective enrichment broth for the detection of Listeria from foods and is not intended for use in the diagnosis of disease or other conditions in humans.Developed to give improved growth rates of Listeria over traditional selective enrichment media, LEE Broth enhances the expression of target antigens for most commercially available immunological test kits/methods whilst maintaining adequate suppression of potential non-target organisms. Selective components are blended into the powder, removing the requirement for supplementation.Compared with more traditional methods, immunological tests, such as ELISA, require relatively high levels of target organisms to achieve a reliable positive result. LEE Broth has been specifically designed to stimulate growth from low numbers to the required high levels within a 24 hour period.For more information visit https://www.neogen.com/solutions/Microbiology/lee-broth
Catalog No. 50-201-5720 Supplier NEOGEN Supplier No. 700004642
Listeria Express Enrichment Broth (LEE Broth) is a selective enrichment broth for the detection of Listeria from foods and is not intended for use in the diagnosis of disease or other conditions in humans.Developed to give improved growth rates of Listeria over traditional selective enrichment media, LEE Broth enhances the expression of target antigens for most commercially available immunological test kits/methods whilst maintaining adequate suppression of potential non-target organisms. Selective components are blended into the powder, removing the requirement for supplementation.Compared with more traditional methods, immunological tests, such as ELISA, require relatively high levels of target organisms to achieve a reliable positive result. LEE Broth has been specifically designed to stimulate growth from low numbers to the required high levels within a 24 hour period.For more information visit https://www.neogen.com/solutions/Microbiology/lee-broth