Learn More
Bachem Cyclo(-Trp-Trp)
SDP
Supplier:  Bachem 4011825.1000
Encompass_Preferred
Cyclo(-Trp-Trp), 20829-55-4, C22H20N4O2, Mr 372.43, 1g. Lee et al. could isolate cyclo(Trp-Trp) from Streptomyces sp. KH29. cWW showed antimicrobial activity against various A. baumannii strains and was highly ative against Staphylococcus aureus TK 784 (MIC 0.8 μg/ml).
Catalog No. 50-194-7461