Learn More
Medchemexpress LLC HY-15777B 500mg , LEE011 (succinate) Ribociclib succinate CAS:1374639-75-4 Purity:98%
SDP
Catalog No. 501492434
Encompass_Preferred
Medchemexpress, HY-15777B 500mg LEE011 (succinate) Ribociclib succinate CAS:1374639-75-4 Formula:C27H36N8O5 IC50: 10/39 nM (CDK4/6) Purity:98% LEE011 succinate is a highly specific CDK4/6 inhibitor with IC 50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2434 Supplier Medchemexpress LLC Supplier No. HY15777B500MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-15777B 500mg LEE011 (succinate) Ribociclib succinate CAS:1374639-75-4 Formula:C27H36N8O5 IC50: 10/39 nM (CDK4/6) Purity:98% LEE011 succinate is a highly specific CDK4/6 inhibitor with IC 50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.