Learn More
Cayman Chemical Leelamine (hydrochloride), Purity: ≥98%, CAS: 16496-99-4, 50 mg
SDP
Catalog No. 502887758
Encompass_Preferred
The hydrochloride of leelamine, an inhibitor of pyruvate dehydrogenase kinase.
Catalog No. 50-288-7758 Supplier Cayman Chemical Supplier No. 1000861450 MG
The hydrochloride of leelamine, an inhibitor of pyruvate dehydrogenase kinase.