Learn More
Medchemexpress LLC Leelamine hydrochloride | 16496-99-4 | 98.1% | 321.93 | 10 MG
SDP
Catalog No. 5000343482
Encompass_Preferred
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and an inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells, functioning independently of androgen receptor status. It also suppresses the transcriptional activity of the androgen receptor, which regulates fatty acid synthesis.
  • Cannabinoid receptor type 1 (CB1) agonist
  • Inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells
  • Suppresses transcriptional activity of androgen receptor
  • Extracted from the bark of pine trees
Catalog No. 50-003-43482 Supplier Medchemexpress LLC Supplier No. HY11002810MG
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and an inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells, functioning independently of androgen receptor status. It also suppresses the transcriptional activity of the androgen receptor, which regulates fatty acid synthesis.
  • Cannabinoid receptor type 1 (CB1) agonist
  • Inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells
  • Suppresses transcriptional activity of androgen receptor
  • Extracted from the bark of pine trees