Learn More
Medchemexpress LLC Leelamine hydrochloride | 16496-99-4 | 98.1% | 321.93 | 100 MG
SDP
Catalog No. 5000343476
Encompass_Preferred
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells, independent of androgen receptor status. It also suppresses the transcriptional activity of the androgen receptor, which regulates fatty acid synthesis.
  • Acts as a cannabinoid receptor type 1 (CB1) agonist.
  • Inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells.
  • Suppresses transcriptional activity of the androgen receptor.
Catalog No. 50-003-43476 Supplier Medchemexpress LLC Supplier No. HY11002810MM/1ML
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells, independent of androgen receptor status. It also suppresses the transcriptional activity of the androgen receptor, which regulates fatty acid synthesis.
  • Acts as a cannabinoid receptor type 1 (CB1) agonist.
  • Inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells.
  • Suppresses transcriptional activity of the androgen receptor.