Learn More
Medchemexpress LLC Leelamine hydrochloride | 16496-99-4 | 98.1% | 321.93 | 25 MG
SDP
Catalog No. 5000347360
Encompass_Preferred
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and an inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells. Its activity is not affected by androgen receptor status, and it suppresses the transcriptional activity of the androgen receptor.
  • Tricyclic diterpene molecule
  • Extracted from the bark of pine trees
  • Cannabinoid receptor type 1 (CB1) agonist
  • Inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells
  • Suppresses transcriptional activity of androgen receptor
  • Appearance: solid
  • Color: white to off-white
  • For research use only
Catalog No. 50-003-47360 Supplier Medchemexpress LLC Supplier No. 25MGHY110028
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and an inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells. Its activity is not affected by androgen receptor status, and it suppresses the transcriptional activity of the androgen receptor.
  • Tricyclic diterpene molecule
  • Extracted from the bark of pine trees
  • Cannabinoid receptor type 1 (CB1) agonist
  • Inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells
  • Suppresses transcriptional activity of androgen receptor
  • Appearance: solid
  • Color: white to off-white
  • For research use only