Learn More
Medchemexpress LLC Leelamine hydrochloride | 16496-99-4 | 98.10% | 321.93 | 1 ML
SDP
Catalog No. 5000344857
Encompass_Preferred
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and an inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells, independent of androgen receptor status. It also suppresses the transcriptional activity of the androgen receptor, which regulates fatty acid synthesis.
  • Acts as a cannabinoid receptor type 1 (CB1) agonist
  • Inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells
  • Suppresses the transcriptional activity of the androgen receptor
  • Extracted from the bark of pine trees
  • Appears as a white to off-white solid
  • Store at -20°C in sealed conditions, away from moisture
  • For storage in solvent, store at -80°C for 6 months, or -20°C for 1 month in sealed conditions, away from moisture
Catalog No. 50-003-44857 Supplier Medchemexpress LLC Supplier No. HY11002850MG
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and an inhibitor of SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells, independent of androgen receptor status. It also suppresses the transcriptional activity of the androgen receptor, which regulates fatty acid synthesis.
  • Acts as a cannabinoid receptor type 1 (CB1) agonist
  • Inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells
  • Suppresses the transcriptional activity of the androgen receptor
  • Extracted from the bark of pine trees
  • Appears as a white to off-white solid
  • Store at -20°C in sealed conditions, away from moisture
  • For storage in solvent, store at -80°C for 6 months, or -20°C for 1 month in sealed conditions, away from moisture