Learn More
Cayman Chemical Leelamine (hydrochloride), Purity: ≥98%, CAS: 16496-99-4, 100 mg
SDP
Catalog No. 502890029
Encompass_Preferred
The hydrochloride of leelamine, an inhibitor of pyruvate dehydrogenase kinase.
Catalog No. 50-289-0029 Supplier Cayman Chemical Supplier No. 10008614100 MG
The hydrochloride of leelamine, an inhibitor of pyruvate dehydrogenase kinase.