Learn More
Medchemexpress LLC Leesggglvqvpggsmk acetate | 1605.76 | 5 MG
SDP
Catalog No. 5000399488
Encompass_Preferred
LEESGGGLVQPGGSMK acetate is a proteolysis peptide and a component of Infliximab. It is utilized for the quantitative analysis of Infliximab, which is a chimeric monoclonal IgG1 antibody that specifically binds to TNF-α. Anti-TNF antibodies have transformed the therapy of TNF-related diseases including Inflammatory Bowel Disease, lupus, ulcerative colitis, ankylosing spondylitis, psoriatic arthritis, and rheumatoid arthritis, by neutralizing TNF activity. These antibodies promote mucosal healing and induce long-term remissions in vivo.
  • Proteolysis peptide
  • Component of Infliximab
  • Used for quantitative analysis of Infliximab
  • Chimeric monoclonal IgG1 antibody that specifically binds to TNF-α
  • Anti-TNF antibodies block the action of TNF alpha
  • Revolutionized therapy for TNF-related diseases
  • Neutralizes TNF activity
  • Promotes mucosal healing
  • Induces long-term remissions in vivo
Catalog No. 50-003-99488 Supplier Medchemexpress LLC Supplier No. HYP3149B5MG
LEESGGGLVQPGGSMK acetate is a proteolysis peptide and a component of Infliximab. It is utilized for the quantitative analysis of Infliximab, which is a chimeric monoclonal IgG1 antibody that specifically binds to TNF-α. Anti-TNF antibodies have transformed the therapy of TNF-related diseases including Inflammatory Bowel Disease, lupus, ulcerative colitis, ankylosing spondylitis, psoriatic arthritis, and rheumatoid arthritis, by neutralizing TNF activity. These antibodies promote mucosal healing and induce long-term remissions in vivo.
  • Proteolysis peptide
  • Component of Infliximab
  • Used for quantitative analysis of Infliximab
  • Chimeric monoclonal IgG1 antibody that specifically binds to TNF-α
  • Anti-TNF antibodies block the action of TNF alpha
  • Revolutionized therapy for TNF-related diseases
  • Neutralizes TNF activity
  • Promotes mucosal healing
  • Induces long-term remissions in vivo