Learn More
Medchemexpress LLC Leesgggvlqpggsmk acetate | 99.0% | 1605.76 | 10 MG
SDP
Catalog No. 5000396613
Encompass_Preferred
LEESGGGLVQPGGSMK acetate is a proteolysis peptide and a component of Infliximab, a chimeric monoclonal IgG1 antibody that specifically binds to TNF-α. This peptide can be used for the quantitative analysis of Infliximab.
  • Can be used for quantitative analysis of infliximab
  • Chimeric monoclonal IgG1 antibody
  • Binds specifically to TNF-α
Catalog No. 50-003-96613 Supplier Medchemexpress LLC Supplier No. HYP3149B10MG
LEESGGGLVQPGGSMK acetate is a proteolysis peptide and a component of Infliximab, a chimeric monoclonal IgG1 antibody that specifically binds to TNF-α. This peptide can be used for the quantitative analysis of Infliximab.
  • Can be used for quantitative analysis of infliximab
  • Chimeric monoclonal IgG1 antibody
  • Binds specifically to TNF-α