Learn More
Aviva Systems Biology Corp LEF1 Antibody - C-terminal region (ARP32403_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32403_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LEF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human LEF1. Alternate names: LEF-1, TCF10, TCF7L3, TCF1ALPHAApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54573