Learn More
Selleck Chemical LLC Lefamulin E4877-5MG
SDP
Catalog No. 5000043532
Encompass_Preferred
Lefamulin (BC-3781) is a pleuromutilin antibiotic inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome It is effective against a range of gram-positive pathogens involved in community-acquired bacterial pneumonia (CABP) including Streptococcus pneumoniae Haemophilus influenzae Mycoplasma pneumoniae Legionella pneumophila Chlamydophila pneumoniae Staphylococcus aureus (including methicillin-resistant and vancomycin-intermediate strains) and vancomycin-resistant Enterococcus faecium
Catalog No. 50-000-43532 Supplier Selleck Chemical LLC Supplier No. E48775MG
Lefamulin (BC-3781) is a pleuromutilin antibiotic inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome It is effective against a range of gram-positive pathogens involved in community-acquired bacterial pneumonia (CABP) including Streptococcus pneumoniae Haemophilus influenzae Mycoplasma pneumoniae Legionella pneumophila Chlamydophila pneumoniae Staphylococcus aureus (including methicillin-resistant and vancomycin-intermediate strains) and vancomycin-resistant Enterococcus faecium