Learn More
Medchemexpress LLC Lefamulin acetate | 1350636-82-6 | 99.7% | 567.78 | 1 ML
SDP
Catalog No. 5000390602
Encompass_Preferred
Lefamulin acetate is an orally active antibiotic that inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome. It also demonstrates anti-inflammatory activity and is utilized in research pertaining to bacterial infections, such as bacterial pneumonia.
  • Inhibits protein synthesis by binding to 50S bacterial ribosome.
  • Exhibits anti-inflammatory activity.
  • Effective against C. trachomatis, N. gonorrhoeae, and M. genitalium.
  • Potent activity against M. pneumoniae strains.
  • Shows antibacterial effect in S. pneumoniae or S. aureus challenged lung infection mice.
  • Reduces BALF neutrophil cell counts and levels of pro-inflammatory cytokines and chemokines in LPS-induced lung neutrophilia mouse model.
Catalog No. 50-003-90602 Supplier Medchemexpress LLC Supplier No. HY16908A10MM/1ML
Lefamulin acetate is an orally active antibiotic that inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome. It also demonstrates anti-inflammatory activity and is utilized in research pertaining to bacterial infections, such as bacterial pneumonia.
  • Inhibits protein synthesis by binding to 50S bacterial ribosome.
  • Exhibits anti-inflammatory activity.
  • Effective against C. trachomatis, N. gonorrhoeae, and M. genitalium.
  • Potent activity against M. pneumoniae strains.
  • Shows antibacterial effect in S. pneumoniae or S. aureus challenged lung infection mice.
  • Reduces BALF neutrophil cell counts and levels of pro-inflammatory cytokines and chemokines in LPS-induced lung neutrophilia mouse model.