Learn More
Medchemexpress LLC Lefamulin (acetate) | 1350636-82-6 | 99.7% | 567.78 | 25 MG
SDP
Catalog No. 5000392655
Encompass_Preferred
Lefamulin (BC-3781) acetate is an orally active antibiotic. It inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome. This product also exhibits anti-inflammatory activity and is used in the research of bacterial infections, such as bacterial pneumonia.
  • Inhibitory activity against *C. trachomatis*, *N. gonorrhoeae*, and *M. genitalium* in vitro.
  • Potent activity against *M. pneumoniae* strains with MIC values of ≤0.008 μg/mL.
  • Shows anti-inflammatory effect on LPS-induced lung neutrophilia mouse models in vivo.
  • Demonstrates antibacterial effect in *S. pneumoniae* or *S. aureus* challenged lung infection mice.
Catalog No. 50-003-92655 Supplier Medchemexpress LLC Supplier No. 25MGHY16908A
Lefamulin (BC-3781) acetate is an orally active antibiotic. It inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome. This product also exhibits anti-inflammatory activity and is used in the research of bacterial infections, such as bacterial pneumonia.
  • Inhibitory activity against *C. trachomatis*, *N. gonorrhoeae*, and *M. genitalium* in vitro.
  • Potent activity against *M. pneumoniae* strains with MIC values of ≤0.008 μg/mL.
  • Shows anti-inflammatory effect on LPS-induced lung neutrophilia mouse models in vivo.
  • Demonstrates antibacterial effect in *S. pneumoniae* or *S. aureus* challenged lung infection mice.