Learn More
Selleck Chemical LLC Lefamulin acetate (Xenleta) S3496-25mg
SDP
Catalog No. 503109298
Encompass_Preferred
Lefamulin acetate (Xenleta BC-3781 acetate) is a pleuromutilin antibiotic for community-acquired bacterial pneumonia (CABP) treatment Lefamulin acetate inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome thus preventing the binding of transfer RNA for peptide transfer
Catalog No. 50-310-9298 Supplier Selleck Chemical LLC Supplier No. S349625MG
Lefamulin acetate (Xenleta BC-3781 acetate) is a pleuromutilin antibiotic for community-acquired bacterial pneumonia (CABP) treatment Lefamulin acetate inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome thus preventing the binding of transfer RNA for peptide transfer