Learn More
Selleck Chemical LLC Lefamulin acetate (Xenleta) S3496-5mg
SDP
Catalog No. 503108770
Encompass_Preferred
Lefamulin acetate (Xenleta BC-3781 acetate) is a pleuromutilin antibiotic for community-acquired bacterial pneumonia (CABP) treatment Lefamulin acetate inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome thus preventing the binding of transfer RNA for peptide transfer
Catalog No. 50-310-8770 Supplier Selleck Chemical LLC Supplier No. S34965MG
Lefamulin acetate (Xenleta BC-3781 acetate) is a pleuromutilin antibiotic for community-acquired bacterial pneumonia (CABP) treatment Lefamulin acetate inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome thus preventing the binding of transfer RNA for peptide transfer