Learn More
TARGETMOL CHEMICALS INC Lefamulin acetate 5MG
SDP
Catalog No. 502876140
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Lefamulin acetate (BC-3781 acetate) (formally BC-3781 acetate) is a novel pleuromutilin antibiotic and is the first to be used for the systemic treatment of bacterial infections in humans. Lefamulin acetate (BC-3781 acetate) inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome, preventing the binding of transfer RNA for peptide transfer.Lefamulin acetate (BC-3781 acetate) is an orally active antibiotic for community-acquired bacterial pneumonia (CABP) treatment. Purity 99.47%
Catalog No. 50-287-6140 Supplier TARGETMOL CHEMICALS INC Supplier No. T118365MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Lefamulin acetate (BC-3781 acetate) (formally BC-3781 acetate) is a novel pleuromutilin antibiotic and is the first to be used for the systemic treatment of bacterial infections in humans. Lefamulin acetate (BC-3781 acetate) inhibits protein synthesis by binding to the peptidyl transferase center of the 50S bacterial ribosome, preventing the binding of transfer RNA for peptide transfer.Lefamulin acetate (BC-3781 acetate) is an orally active antibiotic for community-acquired bacterial pneumonia (CABP) treatment. Purity 99.47%