Learn More
Cayman Chemical Leflunomide, Purity: ≥98%, CAS: 75706-12-6, 25 mg
SDP
Catalog No. 502972906
Encompass_Preferred
A prodrug form of A-771726; inhibits de novo pyrimidine synthesis to regulate T lymphocyte progression through the cell cycle; inhibits proliferation and activation of T cells at 25 and 100 µM, respectively, for naïve and memory CD4+ T cells; reduces and prevents inflammation in a proteoglycan-induced mouse model of rheumatoid arthritis at 35 mg/kg per day
Catalog No. 50-297-2906 Supplier Cayman Chemical Supplier No. 1486025 MG
A prodrug form of A-771726; inhibits de novo pyrimidine synthesis to regulate T lymphocyte progression through the cell cycle; inhibits proliferation and activation of T cells at 25 and 100 µM, respectively, for naïve and memory CD4+ T cells; reduces and prevents inflammation in a proteoglycan-induced mouse model of rheumatoid arthritis at 35 mg/kg per day