Learn More
Selleck Chemical LLC Leflunomide S1247-10mM/1mL
SDP
Catalog No. 503111067
Encompass_Preferred
Leflunomide is a pyrimidine synthesis and protein tyrosine kinase inhibitor belonging to the DMARD used as an immunosuppressant agent The active metabolite of Leflunomide is A77 1726 which inhibits dihydroorotate dehydrogenase (DHODH) Leflunomide is also an agonist of the AhR
Catalog No. 50-311-1067 Supplier Selleck Chemical LLC Supplier No. S124710MM/1ML
Leflunomide is a pyrimidine synthesis and protein tyrosine kinase inhibitor belonging to the DMARD used as an immunosuppressant agent The active metabolite of Leflunomide is A77 1726 which inhibits dihydroorotate dehydrogenase (DHODH) Leflunomide is also an agonist of the AhR