Learn More
Selleck Chemical LLC Leflunomide S1247-1g
SDP
Catalog No. 503111149
Encompass_Preferred
Leflunomide is a pyrimidine synthesis and protein tyrosine kinase inhibitor belonging to the DMARD used as an immunosuppressant agent The active metabolite of Leflunomide is A77 1726 which inhibits dihydroorotate dehydrogenase (DHODH) Leflunomide is also an agonist of the AhR
Catalog No. 50-311-1149 Supplier Selleck Chemical LLC Supplier No. S12471G
Leflunomide is a pyrimidine synthesis and protein tyrosine kinase inhibitor belonging to the DMARD used as an immunosuppressant agent The active metabolite of Leflunomide is A77 1726 which inhibits dihydroorotate dehydrogenase (DHODH) Leflunomide is also an agonist of the AhR