Learn More
TARGETMOL CHEMICALS INC Leflunomide 500MG
SDP
Catalog No. 502877630
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Leflunomide (HWA486) is an immunomodulatory agent used in the therapy of rheumatoid arthritis and psoriatic arthritis. Purity 99.9%
Catalog No. 50-287-7630 Supplier TARGETMOL CHEMICALS INC Supplier No. T1159500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Leflunomide (HWA486) is an immunomodulatory agent used in the therapy of rheumatoid arthritis and psoriatic arthritis. Purity 99.9%