Learn More
Cayman Chemical LeflunomIde-d4 500ug
SDP
Catalog No. 502933996
Encompass_Preferred
An internal standard for the quantification of leflunomide by GC- or LC-MS
Catalog No. 50-293-3996 Supplier Cayman Chemical Supplier No. 26448500 UG
An internal standard for the quantification of leflunomide by GC- or LC-MS