Learn More
Selleck Chemical LLC Leflunomide 100mg 75706-12-6 HWA486
SDP
Supplier:  Selleck Chemical LLC S1247100MG
Encompass_Preferred
Leflunomide (HWA486) is a pyrimidine synthesis and protein tyrosine kinase inhibitor belonging to the DMARD, used as an immunosuppressant agent. The active metabolite of Leflunomide is A77 1726, which inhibits dihydroorotate dehydrogenase (DHODH). Leflunomide is also an agonist of the AhR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-747-6