Learn More
Selleck Chemical LLC Leflunomide 500mg 75706-12-6 HWA486
SDP
Catalog No. 507477
Encompass_Preferred
Leflunomide (HWA486) is a pyrimidine synthesis and protein tyrosine kinase inhibitor belonging to the DMARD, used as an immunosuppressant agent. The active metabolite of Leflunomide is A77 1726, which inhibits dihydroorotate dehydrogenase (DHODH). Leflunomide is also an agonist of the AhR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-747-7 Supplier Selleck Chemical LLC Supplier No. S1247500MG
Leflunomide (HWA486) is a pyrimidine synthesis and protein tyrosine kinase inhibitor belonging to the DMARD, used as an immunosuppressant agent. The active metabolite of Leflunomide is A77 1726, which inhibits dihydroorotate dehydrogenase (DHODH). Leflunomide is also an agonist of the AhR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.