Learn More
BINDER™ Left Access Port (10 mm)
SDP
Catalog No. 08111505 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
08-111-505 Steel
1 options
Catalog No. 08-111-505 Supplier BINDER™ Supplier No. 80121278
Add to Cart
Add to Cart
For Use With: BINDER units series FED
  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 10 mm, left
For Use With (Equipment) BINDER Chambers Series FED
Material Steel
Type Access Port
For Use With (Application) Left
Includes Silicone Plug
Diameter (Metric) Inside 10 mm