Learn More
BINDER™ Access Port with Silicone Plug, Left Access
SDP
Catalog No. 08112017 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
08-112-017 Steel
1 options
Catalog No. 08-112-017 Supplier BINDER™ Supplier No. 80121351
Add to Cart
Add to Cart
BINDER™ Left Access Port is an accessory for use on the left of a variety of chambers and incubators.
  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 125 mm, left
For Use With (Equipment) BINDER Chambers Series MK
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 125 mm