Learn More
Sklar Instruments TC Crile-Wood Left Handed Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1.
SDP
Catalog No. 501437839 Shop All Sklar Instruments Products
Encompass_Preferred
TC Crile-Wood Left Handed Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's TC Left-Handed Crile-Wood Needle Holder is a instrument specially designed for left-handed users to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips, and an overall length of 7 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7839 Supplier Sklar Instruments Supplier No. 219035
TC Crile-Wood Left Handed Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's TC Left-Handed Crile-Wood Needle Holder is a instrument specially designed for left-handed users to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips, and an overall length of 7 inches.