Learn More
Fine Science Tools Surgical Scissors, Sharp-Blunt (Left-Handed), Stainless Steel, 14.5 cm
SDP
Catalog No. NC9348233
Encompass
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Left Handed
Catalog No. NC9348233 Supplier Fine Science Tools Supplier No. 1400814
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Left Handed