Learn More
Aviva Systems Biology Corp LEFTY1 Antibody - N-terminal region (ARP52390_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52390_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LEFTY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LEFTY1. Alternate names: LEFTB, LEFTYBApplicationWB. Species reactivity: Human
Catalog No. 50-014-60725