Learn More
Aviva Systems Biology Corp LEFTY2 Antibody - N-terminal region (ARP42107_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42107_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LEFTY2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LEFTY2. Alternate names: EBAF, LEFTA, LEFTYA, MGC46222, TGFB4ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58110